Learn More
Cole-Parmer™ Nine-Position Stirring Hot Plate
Product Code. 15560492 Shop All Cole-Parmer Products
Click to view available options
Top Plate Material:
Ceramic
Unit Size:
Each
Product Code. 15560492
Brand: Cole-Parmer™ 5145073
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Nine-Position Stirring Hot Plate offers more versatility than single-position units. Each stirring position is individually controlled so you can perform different mixing applications simultaneously.
  • Each stirring position is individually controlled to perform different mixing applications at the same time
    • Front panel controls are positioned to prevent you from touching the hot plate
Specifications
Top Plate Material Ceramic
Voltage 120 V
Includes 6 ft. Cord with Plug
Length (English) 17.5 in.
Height (Metric) 11.43 cm
Temperature (Metric) 100°C to 450°C
Temperature (English) 212°F to 845°F
Height (English) 4.5 in.
Frequency 60 Hz
Width (Metric) 30.48 cm
Amperage 12 amp
Length (Metric) 44.45 cm
Width (English) 12 in.
Type Stirring Hotplate
Max. Sample Load 40 lb.
No. of Positions Multiple
Max. Container Size 0.4 L
Speed 100 to 1500 rpm