Learn More
Nifursemizone, MedChemExpress
Product Code. 16496207
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Product Code. 16496207
Brand: Medchem Express HY101660
MedChemExpress Nifursemizone is an antiprotozoal drug.
Nifursemizone is an antiprotozoal drug.
Specifications
Chemical Name or Material Nifursemizone
Physical Form Solid
CAS 5579-89-5
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C8H10N4O4
For Use With (Application) COVID-19-immunoregulation
Synonym Etafurazone NF161
Solubility Information DMSO : 20.83 mg/mL (92.09 mM; ultrasonic and warming and heat to 60°C)
SMILES O=[N+](C1=CC=C(/C=N/N(C(N)=O)CC)O1)[O-]
Molecular Weight (g/mol) 226.19
Formula Weight 226.19
Percent Purity 99.25%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.