Learn More
Nifuroxazide, TRC
Specifications
Specifications
| Chemical Name or Material | Nifuroxazide |
| CAS | 965-52-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C12 H9 N3 O5 |
| Quantity | 50 mg |
| Synonym | Nifuroxazide, 4-Hydroxybenzoic acid [(5-nitro-2-furanyl)methylene]hydrazide, (5-Nitro-2-furfurylidene)(4-hydroxybenzoyl)hydrazine, 1-(p-Hydroxybenzoyl)-2-(5-nitrofurfurylidene)hydrazine, 27-109RC, 5-Nitro-2-furaldehyde p-hydroxybenzoylhydrazone, 5-Nitro-2-furfurylidene-4-hydroxybenz-hydrazide, Adral, Bacifurane, Diarlidan, Dicoferin, Ercefurol, Ercefuryl, Nifuroxazid, Pentofuryl, RC 27109, RC 30-109, p-Hydroxybenzoic acid (5-nitrofurfurylidene)hydrazide |
| InChI Formula | InChI=1S/C12H9N3O5/c16-9-3-1-8(2-4-9)12(17)14-13-7-10-5-6-11(20-10)15(18)19/h1-7,16H,(H,14,17)/b13-7+ |
| SMILES | Oc1ccc(cc1)C(=O)N\N=C\c2oc(cc2)[N+](=O)[O-] |
| IUPAC Name | 4-hydroxy-N-[(E)-(5-nitrofuran-2-yl)methylideneamino]benzamide |
| Molecular Weight (g/mol) | 275.22 |
| Show More |