missing translation for 'onlineSavingsMsg'
Learn More

Nifuroxazide, MedChemExpress

Product Code. p-200099033
Encompass_Preferred
Click to view available options
Quantity:
200 mg
500 mg
Unit Size:
200mg
500mg
This item is not returnable. View return policy

Product Code. 16461456

Brand: Medchem Express HYB1436

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nifuroxazide is an effective inhibitor of STAT3, also exerts potent anti-tumor and anti-metastasis activity.

Nifuroxazide is an effective inhibitor of STAT3, also exerts potent anti-tumor and anti-metastasis activity.

Specifications

Chemical Name or Material Nifuroxazide
CAS 965-52-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C12H9N3O5
Quantity 200 mg
Solubility Information DMSO : ≥ 155 mg/mL (563.19 mM)
SMILES O=C(N/N=C/C1=CC=C([N+]([O-])=O)O1)C2=CC=C(O)C=C2
Molecular Weight (g/mol) 275.22
Formula Weight 275.22
Percent Purity 98.51%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.