Learn More
Niflumic Acid, LoGiCal
Product Code. 30460304
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30460304 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30460304 Supplier LoGiCal Supplier No. LGCFOR0850.00
 Restricted Item
Estimated Shipment: 30-11-2026
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Niflumic Acid
CAS 4394-00-7
Analyte or Component Names Niflumic Acid
Shipping Condition Room Temperature
Molecular Formula C13 H9 F3 N2 O2
Certifications/Compliance ISO 9001
Synonym Niflumic Acid, 2-[[3-(Trifluoromethyl)phenyl]amino]-3-pyridinecarboxylic acid, 2-[3-(Trifluoromethyl)anilino]nicotinic acid, UP-83, Actol, Forenol, Landruma, Nifluril, 2-(alpha,alpha,alpha-Trifluoro-m-toluidino)nicotinic acid
InChI Formula InChI=1S/C13H9F3N2O2/c14-13(15,16)8-3-1-4-9(7-8)18-11-10(12(19)20)5-2-6-17-11/h1-7H,(H,17,18)(H,19,20)
Solution Type Neat
SMILES OC(=O)c1cccnc1Nc2cccc(c2)C(F)(F)F
IUPAC Name 2-[3-(trifluoromethyl)anilino]pyridine-3-carboxylic acid
Industry Type Clinical Testing
Molecular Weight (g/mol) 282.22
Quantity 10 mg
Formula Weight 282.0616 g/mol
Show More Show Less
Product Title
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.