missing translation for 'onlineSavingsMsg'
Learn More

Nicotinamide Adenine Dinucleotide Free Acid, Millipore™

Product Code. 10726121
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10726121 1 g 1g
10192930 5 g 5g
10187830 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10726121 Supplier Millipore Supplier No. 1245420001
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For biochemistry NAD

We are committed to bringing you Greener Alternative Products that adhere to one or more of the 12 Principles of Green Chemistry. Beta-Nicotinamide adenine dinucleotide is a bio-derived redox cofactor that works together with enzymes as part of a biocatalyst system. It can be efficiently recycled in many reactions, enabling selective oxidations while reducing waste and supporting greener, more sustainable chemical processes.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53-84-9

Specifications

Molecular Weight (g/mol) 663.43
Product Type NADP-Na2
Quantity 1 g
Grade MQ200 - for Research, and Non-Regulated Industrial Applications
Synonym Nicotinamide adenine dinucleotide free acid, NAD free acid, DPN, Nicotinamide adenine dinucleotide free acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.