Learn More
Swann-Morton™ Nickel/Stainless Steel Post Mortem Handle
Product Code. 12932555
Change view
Click to view available options
Blade Material:
Nickel-plated Steel
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Blade Material Unit Size
12932555 Nickel-plated Steel Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12932555 Supplier Swann-Morton™ Supplier No. 2650
  • ONLINE EXCLUSIVE PRICE
    712.00 NOK
    639.53 NOK / Pack of 1
    Save 72.47 NOK
    10% Off
View all promotions
Estimated Shipment: 24-11-2026
Specifications
Blade Material Nickel-plated Steel
Product Type Postmortem Handle
For Use With (Equipment) Traditional PM40 and PM40 Bull Nose Blade