Description
Designed for precise sampling of powders or salts. Nickel/stainless steel spatulas come in a variety of end styles.
- Nickel/SS Lab Spatulas
- Available with a glazed or mirror finish
- Easy to clean
- Multiple sizes and tyles to fit a variety of laboratory needs
- Pack of 3
Specifications
Specifications
| Quantity | 3 Pk. |
| Shape | Round End |
| Product Type | Laboratory Spatula |
| Length (English) | 7.125 in. |
| Material | Stainless Steel (304), Nickel |
| Handle Type | Round End |
| Length (Metric) | 18.1 cm |