missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) Sulfate Hexahydrate, Supelco™

Product Code. 15867520
Change view
Click to view available options
Quantity:
1 kg
50 kg
Unit Size:
1kg
50kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15867520 50 kg 50kg
15857520 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15867520 Supplier Supelco Supplier No. 1067269050
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

EMPLURA™

EMPLURA™

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-97-0

Specifications

Quantity 50 kg
Grade Research
Density 2.07 g/cm3 at 20°C
Chemical Name or Material Nickel(II) Sulfate Hexahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.