Learn More
Bochem™ Nickel Crucibles
Specifications
Specifications
| Material | Nickel |
| Capacity (Metric) | 130 mL |
| Diameter (Metric) Outer | 60mm |
| Height (Metric) | 60mm |
| Product Type | Crucible |