Learn More
Thermo Scientific Chemicals Nickel Cover for Crucible 35925, 50ml
Product Code. 15451377
Change view
Click to view available options
Capacity (Metric):
5 mL
20 mL
25 mL
30 mL
35 mL
45 mL
50 mL
55 mL
75 mL
100 mL
250 mL
500 mL
1000 mL
Description:
For 35883 crucible
For 35889 crucible
For 35904 crucible
For 35906 crucible
For 35925 crucible
For 35927 crucible
For 35946 crucible
For 35969 crucible
For 35991 crucible
For 36019 crucible
For 36034 crucible
For 36040 crucible
For 36053 crucible
For 36057 crucible
For 36071 crucible
For 36084 crucible
Unit Size:
Each
Product Code. Capacity (Metric) Description unitSize
15451377 100 mL For 36057 crucible Each
15461357 1000 mL For 36034 crucible Each
15411367 500 mL For 36040 crucible Each
15471367 250 mL For 36084 crucible Each
15491367 250 mL For 36019 crucible Each
15401387 100 mL For 36053 crucible Each
15431397 75 mL For 35946 crucible Each
15441397 20 mL For 35883 crucible Each
15491397 5 mL For 35889 crucible Each
15421407 50 mL For 35925 crucible Each
15441407 45 mL For 35969 crucible Each
15471407 55 mL For 36071 crucible Each
15411417 20 mL For 35906 crucible Each
15461417 30 mL For 35904 crucible Each
15401437 25 mL For 35927 crucible Each
15411447 35 mL For 35991 crucible Each
16 options
Product Code. 15451377
Brand: Thermo Scientific Chemicals 035942.KT
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Description For 36057 crucible
Material Nickel
Capacity (Metric) 100 mL
Product Type Nickel Crucible Cover