missing translation for 'onlineSavingsMsg'
Learn More

Nickel Chromium pieces, 1.0mm (0.04 in.) dia. x 3.18mm (0.125 in.) length, 99% (metals basis), Si nominal 1.5%, Thermo Scientific™

Product Code. p-7051092
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. Quantity unitSize
11398388 5 g 5g
11388388 25 g 25g
11378388 100 g 100g
3 options
This item is not returnable. View return policy

Product Code. 11398388

Brand: Thermo Scientific Alfa Aesar 040437.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High resistivity material for electronics industry

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11106-97-1
Molecular Formula CrNi
Molecular Weight (g/mol) 110.69
MDL Number MFCD00673110
InChI Key VNNRSPGTAMTISX-UHFFFAOYSA-N
Synonym nickel chromium, nickel-chromium, nicr, nickel chromium lump, chromium, compd. with nickel 1:1, chromium nickel, acmc-1byoe, nickel chromium powder,-140+325 mesh, nickel chromium wire, 1.0mm 0.04in dia, nickel chromium wire, 0.25mm 0.01in dia
PubChem CID 11320920
IUPAC Name chromium;nickel
SMILES [Cr].[Ni]

Specifications

Avg. Mol. Wt. or Mol. Wt. Range Ni:Cr; 80:20 wt%
Quantity 5 g
Odor Odorless
Weight ≈0.21g/piece
Solubility Information Insoluble in water.
Percent Purity 99%
Concentration or Composition (by Analyte or Components) Si nomin.al 1.5%
Assay (metals basis)
Chemical Name or Material Nickel Chromium pieces, 1.0mm (0.04 in.) dia. x 3.18mm (0.125 in.) length
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.