missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NGFR Monoclonal Antibody (OTI8A7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812986S
This item is not returnable.
View return policy
Description
NGFR Monoclonal antibody specifically detects NGFR in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
| NGFR | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P07174, P08138, Q9Z0W1 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 18053, 24596, 4804 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry | |
| OTI8A7 | |
| Unconjugated | |
| Ngfr | |
| CD271; CD271 antigen; Gp80-LNGFR; LNGFR; low affinity nerve growth factor receptor; low affinity neurotrophin receptor p75NTR; LOW QUALITY PROTEIN: tumor necrosis factor receptor superfamily member 16; low-affinity nerve growth factor receptor; nerve growth factor receptor; nerve growth factor receptor (TNFR superfamily, member 16); nerve growth factor receptor, fast; NGF receptor; NGFR; NGR; p75; p-75; p75 ICD; p75 neurotrophin receptor; p75 ntr; p75(NTR); p75NGFR; p75NTR; RNNGFRR; TNFR superfamily, member 16; Tnfrsf16; TNR16; Tumor necrosis factor receptor superfamily member 16 | |
| Full length human recombinant protein of human Ngfr (NP_002498) produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction