Learn More
LNI Swissgas NGA CASTORE XL iQ QSight DS
Product Code. 17851989 Shop All LNI Swissgas Products
Change view
Click to view available options
Voltage:
100/115/220/230 V
Unit Size:
Each
Product Code. Voltage unitSize
17851989 100/115/220/230 V Each
1 options
Product Code. 17851989
Brand: LNI Swissgas 6920.70.24
Nitrogen and dry air generator for PE QSight double source.
  • With integrated compressor.
Specifications
Voltage 100/115/220/230 V
Type Nitrogen Gas Generator