missing translation for 'onlineSavingsMsg'
Learn More

NFIL3 Antibody (714401), mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18549397 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18549397 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18549397 Supplier Novus Biologicals Supplier No. FAB8570MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NFIL3 Monoclonal antibody specifically detects NFIL3 in Human samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFIL3
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 714401
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias E4BP4IL3BP1, Interleukin-3 promoter transcriptional activator, Interleukin-3-binding protein 1, NF-IL3A, NFIL3AE4 promoter-binding protein 4, nuclear factor interleukin 3 regulated protein, nuclear factor interleukin-3-regulated protein, nuclear factor, interleukin 3 regulated, Transcriptional activator NF-IL3A
Host Species Mouse
Immunogen E. coli-derived recombinant human NFIL3/E4BP4, Lys140-Gly462, Accession # Q16649
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4783
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.