Learn More
NFATC3 Mouse anti-Human, Unconjugated, Clone: 1C1H8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66924-1-IG-20UL
 View more versions of this product
Product Code. 16849225
  • 1710.33 NOK / 20µL
Description
The NFATC3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NFATC3. This antibody recognizes human antigen. The NFATC3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
NFATC3
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot
1C1H8
Unconjugated
20 μL
4775
IgG1