missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NF-H Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21471-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody recognize NEFH only.
Involved in the maintece of neuronal caliber, neurofilaments are the intermediate filament proteins found specifically in neurons, and are composed predomitly of three major proteins called NF-L, NF-M and NF-H. Like most other intermediate filament proteins (IFPs), the expression of the different neuronal IFPs is both tissue-specific and developmentally regulated. NF-H has an important role in mature axons and is involved in amyotrophic lateral sclerosis (ALS).Specifications
| NF-H | |
| Polyclonal | |
| Unconjugated | |
| NEFH | |
| Nefh | |
| NF-H Fusion Protein Ag15249 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P12036, P16884, P19246 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 24587, 380684, 4744 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction