Learn More
Emma Safety Footwear New York Safety Shoes
Product Code. 15908441
Change view
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15908441 47 1 pair
15928431 39 1 pair
15938431 40 1 pair
15948431 41 1 pair
15958431 42 1 pair
15968431 43 1 pair
15978431 44 1 pair
15988431 45 1 pair
15998431 46 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 15908441 Supplier Emma Safety Footwear Supplier No. MM43264747
Only null left
Add to basket
Add to basket
Light and comfortable with knitted textile upper and Nano Carbon Composite protective nose. Ideal for workplaces where safety and long-lasting comfort are top priorities. Integrated lacing prevents pressure points from hooks and eyes and the Hydro-Tec™ lining provides optimum breathability.
  • S1P-Safety shoe with sole plate for dry work
  • Knitted Textile
  • Polyurethane coated
  • Hydro-Tec™ Antibacterial
  • Metal-free
  • Chrome-free lining
  • ESD certified
Specifications
Certifications/Compliance EN ISO 20345:2011
Closure Type Laces
Color Gray, Black
Design New York
Liner Material Hydro-Tec™ Antibacterial
Material Textile
Size 47
Toe Material Composite