Learn More
Eppendorf™ Subplatform for Model C76 Water Bath Shaker
Click to view available options
For Use With (Equipment):
Model C76 water bath shaker
Unit Size:
Each
Description
Use as mounting base for two half-size platforms or four test-tube racks.
Specifications
Specifications
| For Use With (Equipment) | Model C76 water bath shaker |
| Type | Sub Platform |