Learn More
Eppendorf™ Scientific Test Tube Racks
Adjustable angle and tube height
Brand: Eppendorf™ M1289-0002
Additional Details : Weight : 2.49480kg
Description
- Inserts to help retain tubes and prevent them from spinning in the rack
- Finger-tight knobs to quickly adjust angle, or release the rack from the base plate, for easy transport to the bench
- Stainless-steel
Specifications
Test Tube Rack | |
12 to 15 mm | |
Two adjustable bars and non-skid rubber mat | |
Small |
Stainless Steel | |
Innova 43/43R Incubator shaker | |
34 |