Learn More
Eppendorf™ New Brunswick Scientific™ CO2 back-up
Product Code. 11622834
Click to view available options
:
For use with: CO2 back-up
Product Code. 11622834
Brand: Eppendorf™ U90430004
Order Info
ForUseWithEquipment
TRUSTED_SUSTAINABILITY
Specifications
Type CO2 Backup System
Color Innova and Green
For Use With (Equipment) CO2 back-up
Voltage 230 V