missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neutrophil Elastase/ELA2 Antibody (950317R) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB91671RJF635
This item is not returnable.
View return policy
Description
Neutrophil Elastase/ELA2 Monoclonal antibody specifically detects Neutrophil Elastase/ELA2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Flow Cytometry (Intracellular Staining),Immunocytochemistry,CyTOF
Specifications
| Neutrophil Elastase/ELA2 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| Bone marrow serine protease, EC 3.4.21, EC 3.4.21.37, ELA2granulocyte-derived elastase, elastase 2, neutrophil, elastase, neutrophil expressed, Elastase-2, GE, HLEelastase-2, HNE, Human leukocyte elastase, leukocyte elastase, medullasin, NE, neutrophil elastase, PMN elastase, PMN-E, polymorphonuclear elastase, SCN1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Neutrophil Elastase/ELA2, Met1-Asn252, Accession # P08246 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry, Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
| 950317R | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cancer, Neuroscience | |
| 1991 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering