missing translation for 'onlineSavingsMsg'
Learn More

Neutral Red (C.I. 50040), Supelco™

Product Code. 15896980
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15896980 100 g 100g
15874070 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15896980 Supplier Supelco Supplier No. 1013690100
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Indicator and for microbiology

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 553-24-2

Specifications

MDL Number MFCD00012651
Synonym Neutral red (C.I. 50040), Neutral red (C.I. 50040)
Quantity 100 g
Molecular Weight (g/mol) 288.78
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.