missing translation for 'onlineSavingsMsg'
Learn More

NeutraKine™ IL-23 p40 Mouse anti-Human, Unconjugated, Clone: 2A9H6, Proteintech

Product Code. 16860416 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
Product Code. Quantity unitSize
16860416 25 μg 25µg
16850416 100 μg 100µg
2 options
This item is not returnable. View return policy

Product Code. 16860416

Brand: Proteintech 690061IG25UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The NeutraKine™ IL-23 p40 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NeutraKine™ IL-23 p40. This antibody recognizes human antigen. The NeutraKine™ IL-23 p40 antibody has been validated for the following applications: Neutralization, ELISA analysis.

Specifications

Antigen NeutraKine™ IL-23 p40
Applications Neutralization
Classification Monoclonal
Clone 2A9H6
Conjugate Unconjugated
Host Species Mouse
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 51561
Target Species Human
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.