missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NeutraKine FGF-4 Mouse anti-Human, Unconjugated, Clone: 1A2D8, Proteintech
Shop All Proteintech ProductsDescription
This antibody is used to Block/Neutralize FGF-4 protein.
Specifications
Specifications
| Antigen | NeutraKine FGF-4 |
| Applications | Neutralization, ELISA |
| Classification | Monoclonal |
| Clone | 1A2D8 |
| Concentration | 1000 μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Quantity | 100 μg |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 2249 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?