Learn More
Neuroserpin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12558-1-AP-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16850324
  • 1415.13 NOK / 20µL
Estimated Shipment: 17-10-2023
Description
This gene encodes a member of the serpin superfamily of serine proteinase inhibitors. The protein is primarily secreted by axons in the brain, and preferentially reacts with and inhibits tissue-type plasminogen activator. It is thought to play a role in the regulation of axonal growth and the development of synaptic plasticity. Mutations in this gene result in familial encephalopathy with neuroserpin inclusion bodies (FENIB), which is a domitly inherited form of familial encephalopathy and epilepsy characterized by the accumulation of mutant neuroserpin polymers. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Specifications
Specifications
Neuroserpin
Polyclonal
Unconjugated
SERPINI1
neuroserpin, Peptidase inhibitor 12, PI 12, PI12, Serpin I1, SERPINI1
Rabbit
Antigen Affinity Chromatography
RUO
116459, 20713, 5274
-20°C
Liquid
Western Blot
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O35684, Q99574, Q9JLD2
SERPINI1
Neuroserpin Fusion Protein Ag3275
20 μL
Primary
Rat, Human, Mouse