missing translation for 'onlineSavingsMsg'
Learn More

Neuronal Pentraxin 1 Antibody (OTI5G9), PerCP, Novus Biologicals™

Product Code. 30034303 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
Product Code. Quantity unitSize
30034303 0.1 mL 0.10mL
1 options
This item is not returnable. View return policy

Product Code. 30034303

Brand: Novus Biologicals NBP272965PCP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Neuronal Pentraxin 1 Monoclonal antibody specifically detects Neuronal Pentraxin 1 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neuronal Pentraxin 1
Applications Western Blot
Classification Monoclonal
Clone OTI5G9
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias DKFZp686J2446, MGC105123, neuronal pentraxin INP1NP-I, neuronal pentraxin-1
Host Species Mouse
Immunogen Full length human recombinant protein of human NPTX1 (NP_002513) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4884
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.