missing translation for 'onlineSavingsMsg'
Learn More
Learn More
neuromedin B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10888-1-AP-20UL
This item is not returnable.
View return policy
Description
NMB stimulates smooth muscle contraction in a manner similar to that of bombesin.Specifications
| neuromedin B | |
| Polyclonal | |
| Unconjugated | |
| NMB | |
| neuromedin B, NMB | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4828, 499194, 68039 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P08949, Q9CR53 | |
| NMB | |
| neuromedin B Fusion Protein Ag0842 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction