Learn More
neuromedin B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10888-1-AP-150UL
 View more versions of this product
Product Code. 16837573
  • 4836.10 NOK / 150µL
Description
NMB stimulates smooth muscle contraction in a manner similar to that of bombesin.
Specifications
Specifications
neuromedin B
Polyclonal
Unconjugated
NMB
neuromedin B, NMB
Rabbit
Antigen Affinity Chromatography
RUO
4828, 499194, 68039
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P08949, Q9CR53
NMB
neuromedin B Fusion Protein Ag0842
150 μL
Primary
Human, Mouse, Rat