Learn More
NEUROD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12081-1-AP-150UL
This item is not returnable.
View return policy
Description
NeuroD1 encodes a member of the NeuroD family of basic helix-loop-helix (bHLH) transcription factors. The protein forms heterodimers with other bHLH proteins and activates transcription of genes that contain a specific DNA sequence known as the E-box. It regulates expression of the insulin gene, and mutations in this gene result in type II diabetes mellitus.Specifications
| NEUROD1 | |
| Polyclonal | |
| Unconjugated | |
| NEUROD1 | |
| BETA2, BHF 1, bHLHa3, NEUROD, NEUROD1, neurogenic differentiation 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18012, 29458, 4760 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.47 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q13562, Q60867, Q64289 | |
| Neurod1 | |
| NEUROD1 Fusion Protein Ag2713 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |