missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEUROD1 Mouse anti-Human, Clone: 3E10F1, Proteintech
Mouse Monoclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | NEUROD1 |
---|---|
Clone | 3E10F1 |
Concentration | 1 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16888805
|
Proteintech
66691-1-IG-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16878805
|
Proteintech
66691-1-IG-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
NeuroD1 encodes a member of the NeuroD family of basic helix-loop-helix (bHLH) transcription factors. The protein forms heterodimers with other bHLH proteins and activates transcription of genes that contain a specific DNA sequence known as the E-box. It regulates expression of the insulin gene, and mutations in this gene result in type II diabetes mellitus.Specifications
NEUROD1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
BETA2, BHF 1, bHLHa3, NEUROD, NEUROD1, neurogenic differentiation 1 | |
NEUROD1 | |
IgG1 | |
Protein G | |
Antibody |
3E10F1 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
Q13562 | |
4760 | |
NEUROD1 Fusion Protein Ag27606 | |
Primary | |
-20°C | |
NEUROD1 |