Learn More
NeuN Mouse anti-Human, Unconjugated, Clone: 3A4C1, Proteintech
Product Code. 16809085 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16809085 20 μL 20µL
16899075 150 μL 150µL
2 options
Product Code. 16809085
Brand: Proteintech 668361IG20UL
Mouse Monoclonal Antibody
The NeuN antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NeuN. This antibody recognizes human, mouse, rat antigen. The NeuN antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
Antigen NeuN
Applications Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 3A4C1
Concentration 1000 μg/mL
Conjugate Unconjugated
Host Species Mouse
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 146713
Target Species Human, Rat, Mouse
Isotype IgG1