Learn More
NeuN Mouse anti-Human, Unconjugated, Clone: 3A4C1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66836-1-IG-150UL
Description
The NeuN antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NeuN. This antibody recognizes human, mouse, rat antigen. The NeuN antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
| NeuN | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Immunohistochemistry, Immunofluorescence | |
| 3A4C1 | |
| Unconjugated | |
| 150 μL | |
| 146713 | |
| IgG1 |