missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NeuN Mouse anti-Human, Mouse, Rat, CoraLite 555, Clone: 3A4C1, Proteintech
Mouse Monoclonal Antibody
1651.35 NOK - 5307.92 NOK
Specifications
| Antigen | NeuN |
|---|---|
| Clone | 3A4C1 |
| Applications | Immunofluorescence, Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | CoraLite 555 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17721369
|
Proteintech
CL555-66836-20UL |
20 μL |
1651.35 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17609916
|
Proteintech
CL555-66836-100UL |
100 μL |
5307.92 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NeuN Monoclonal antibody apecifically detetects NeuN antigen in Human, Mouse, Rat Samples. It is validated for Immunofluorescence, Flow Cytometry.Specifications
| NeuN | |
| Immunofluorescence, Flow Cytometry | |
| CoraLite 555 | |
| Mouse | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| 146713 | |
| NeuN fusion protein Ag28016 | |
| Primary | |
| Store at -20°C. Avoid exposure to light. |
| 3A4C1 | |
| Monoclonal | |
| Liquid | |
| Human, Mouse, Rat | |
| NM_001082575 | |
| hCG_1776007 | |
| IgG1 | |
| Protein G purification |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title