missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NeuN Mouse anti-Human, Mouse, Rat, CoraLite 555, Clone: 3A4C1, Proteintech
Shop All Proteintech ProductsDescription
NeuN Monoclonal antibody apecifically detetects NeuN antigen in Human, Mouse, Rat Samples. It is validated for Immunofluorescence, Flow Cytometry.
Specifications
Specifications
| Antigen | NeuN |
| Applications | Immunofluorescence, Flow Cytometry |
| Classification | Monoclonal |
| Clone | 3A4C1 |
| Concentration | 1000 ug/mL |
| Conjugate | CoraLite 555 |
| Formulation | PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. |
| Gene Accession No. | NM_001082575 |
| Gene Symbols | hCG_1776007 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?