Learn More
NET1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28180-1-AP-150UL
Description
The NET1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NET1. This antibody recognizes human, mouse antigen. The NET1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| NET1 | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 10276 | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 467 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse |