Learn More
Cytiva Net Ring
Shop All Cytiva Products
missing translation for 'orderingAttributeHoverText'
For Use With:
For 26 mm HiScale Columns I.D.
For 50 mm HiScale Columns I.D.
missing translation for 'unitSize'
Each
Pack of 5
Specifications
Specifications
| Type | Net Ring |
| Packaging Quantity | 5 Pack |
| For Use With | For 50 mm HiScale Columns I.D. |