Learn More
Nepsilon-Boc-L-lysine methyl ester hydrochloride, 98%
Product Code. 15410569
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 15410569
Brand: Thermo Scientific Alfa Aesar H63727.03
Request Bulk or Custom Format
CAS: 2389-48-2 | C12H25ClN2O4 | 296.79 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in methanol.
Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2389-48-2
Molecular Formula C12H25ClN2O4
Molecular Weight (g/mol) 296.79
MDL Number MFCD00076959
InChI Key NANRHOPPXCBHGI-MVRPGTNWNA-N
Synonym h-lys boc-ome.hcl, h-lys boc-ome hcl, h-lys boc-ome hydrochloride, lys boc-ome.hcl, s-methyl 2-amino-6-tert-butoxycarbonyl amino hexanoate hydrochloride, methyl 2s-2-amino-6-tert-butoxycarbonyl amino hexanoate hydrochloride, pubchem18976, h-lys boc-ome?cl, lys boc-ome. hcl, h-lys boc-ome??hcl
PubChem CID 16218818
IUPAC Name methyl (2S)-2-amino-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoate;hydrochloride
SMILES Cl.COC(=O)[C@@H](N)CCCCNC(=O)OC(C)(C)C
Specifications
Beilstein 5173577
Solubility Information Soluble in methanol.
Quantity 1 g
Formula Weight 296.79
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Nepsilon-Boc-L-lysine methyl ester hydrochloride