Learn More
Nepsilon-Acetyl-L-lysine, MedChemExpress
Product Code. 16425586
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16425586 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16425586 Supplier Medchem Express Supplier No. HY113426
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress Nepsilon-Acetyl-L-lysine is a derivative of the amino acid lysine.
Nepsilon-Acetyl-L-lysine is a derivative of the amino acid lysine.
Specifications
Chemical Name or Material Nepsilon-Acetyl-L-lysine
CAS 692-04-6
Color White
Purity Grade Notes Research
Molecular Formula C8H16N2O3
Solubility Information H2O : 50 mg/mL (265.65 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (5.31 mM; ultrasonic and warming and heat to 60°C)
SMILES N[C@@H](CCCCNC(C)=O)C(O)=O
Molecular Weight (g/mol) 188.22
Quantity 100 mg
Formula Weight 188.22
Percent Purity 97.0%
Grade Research
Physical Form Solid