Description
- Permits intermittent handling in hot liquids up to 180°C and handling in cold temperatures down to -25°C.
- Comfortable anatomical glove shape which follows the contours of the hand at rest.
- Improves dexterity and reduces hand fatigue.
- Good resistance to all mechanical hazards.
- Neoprene Coating.
- Smooth finish grip design.
Specifications
Specifications
| Back Material | Neoprene |
| Certifications/Compliance | Standard CATEGORY III, EN 374, EN 388 : 3121, ANSI, FDA 21 CFR 177.2600 |
| Cuff Material | Neoprene |
| Coating Material | Neoprene |
| Product Type | Immersion Gloves |
| Size | 10 |
| Style | 09-022 |
| Temperature (Metric) Max | 180°C |
| Glove Cuff Style | Gauntlet |
| Lining | Cotton Jersey |
| Show More |