Learn More
Invitrogen™ Neon™ NxT Electroporation System 1-Channel Tubes
Product Code. 30245030
Change view
Click to view available options
Quantity:
24 Tubes
96 tubes
Unit Size:
Pack of 24
Pack of 96
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30245030 24 Tubes Pack of 24
18065321 96 tubes Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30245030 Supplier Invitrogen™ Supplier No. NT24
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
Neon NxT Electroporation System 1-Channel Tubes are designed specifically for use with the Neon NxT Electroporation System with 1-Channel Pipette.
Neon NxT Electroporation System 1-Channel Tubes are designed specifically for use with the Neon NxT Electroporation System with 1-Channel Pipette (Cat. Nos. NEON1S, NEON1SK, NEON18S, NEON18SK). Each Neon NxT Electroporation System Kit for 25 x 2 reactions (Cat. Nos. N1025 and N10025) comes with eight 1-channel tubes, and each Neon NxT Electroporation System Kit with 1-Channel Tubes for 96 x 2 reactions (Cat. Nos. N1096 and N10096) comes with 32 1-channel tubes. This pack of additional tubes is offered for those wishing to use single tubes per reaction depending on their experimental setup or for reactions with the 1-channel pipette when predominately using the 8-channel pipette and utilizing kits that contain the 8-channel tubes.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Includes 24 tubes, use at room temperature under normal laboratory conditions.
Refer to the Neon™ NxT Electroporation System user manual for protocol instructions.
Includes Includes 24 tubes, use at room temperature under normal laboratory conditions. Refer to the Neon™ NxT Electroporation System user manual for protocol instructions.
Quantity 24 Tubes
Cell Type Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format Electroporation Pipette Tip
For Use With (Equipment) Neon™ NxT Electroporation System
Transfection Technique Electroporation
For Use With (Application) Electroporation, Transfection