Learn More
Invitrogen™ Neon™ NxT Electroporation System 1-Channel Tubes
Product Code. 18065321
Click to view available options
Quantity:
24 Tubes
96 tubes
Unit Size:
Pack of 24
Pack of 96
Product Code. 18065321
Brand: Invitrogen™ NT96
Neon NxT Electroporation System 1-Channel Tubes are designed specifically for use with the Neon NxT Electroporation System with 1-Channel Pipette.
Neon NxT Electroporation System 1-Channel Tubes are designed specifically for use with the Neon NxT Electroporation System with 1-Channel Pipette (Cat. Nos. NEON1S, NEON1SK, NEON18S, NEON18SK). Each Neon NxT Electroporation System Kit for 25 x 2 reactions (Cat. Nos. N1025 and N10025) comes with eight 1-channel tubes, and each Neon NxT Electroporation System Kit with 1-Channel Tubes for 96 x 2 reactions (Cat. Nos. N1096 and N10096) comes with 32 1-channel tubes. This pack of additional tubes is offered for those wishing to use single tubes per reaction depending on their experimental setup or for reactions with the 1-channel pipette when predominately using the 8-channel pipette and utilizing kits that contain the 8-channel tubes.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 96 tubes
Cell Type Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format Electroporation Pipette Tip
For Use With (Equipment) Neon™ NxT Electroporation System
Transfection Technique Electroporation
For Use With (Application) Electroporation, Transfection