Learn More
Neomycin Sulfate, Phytopure™, MP Biomedicals™
Product Code. 17600907 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
Product Code. Quantity unitSize
17600907 10 g 10g
17610907 25 g 25g
2 options
Product Code. 17600907
Brand: MP Biomedicals 0219453310
Synonym: Neomycin B Sulfate, Bykomycin, Endomixin, Fraquinol, Myacine, Neosulf, Neomix
Neomycin sulfate is an aminoglycoside antibiotic active against gram-negative bacteria. It is commonly used in many topical medications, such as creams, ointments, and eyedrops. It binds to duplex RNA with high affinity and inhibits protein biosynthesis. Neomycin sulfate also acts as a calcium channel blocker. Usage Statement: Unless specified otherwise, MP Biomedical™ products are for research or further manufacturing use only, not for direct human use.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1405-10-3
Specifications
Physical Form Powder
Color White
Quantity 10 g
compliance-icons compliance-icons
Product Identifier
  • Neomycin Sulfate, Cell Culture Reagent
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P321-Specific treatment (see on this label).
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Neomycin Sulfate