Learn More
Dr.Weigert™ Neodisher LaboClean FLA
Product Code. 11758373
Change view
Click to view available options
Volume (Metric):
5 L
10 L
Unit Size:
10 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11758373 10 L 10 L
11768373 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11758373 Supplier Dr.Weigert™ Supplier No. 411230
  • ONLINE EXCLUSIVE PRICE
    1410.00 NOK
    1080.00 NOK / 10 L
    Save 330.00 NOK
    23% Off
 Restricted Item
Estimated Shipment: 18-11-2026
Neodisher LaboClean FLA - Alkaline detergent, liquid concentrate for the automated cleaning of commonly used laboratory utensils (in medical, biological, microbiological, chemical, food, pharmaceutical, ... ). Removes stubborn oily residues and other organic residues, e.g. fats, oils, starch, residues of felt-tip pens
  • Alkaline detergent, liquid concentrate
  • neodisher LaboClean FLA is used for cleaning in washer disinfection
  • Free of phosphates, surfactants and active chlorine
  • Removes organic residues, especially suitable for cleaning animal cages for mice or rats made of polysulfone, polycarbonate, polyetherimide, polyphenylsulfone or stainless steel
  • Dosage of between 2-6 mL/L
Specifications
Product Type Neodisher FLA
Form Liquid
pH 12.3 to 12.8
Sterility Non-sterile
Composition Polycarboxylates, potassium hydroxide
Concentration 1 to 6 mL/L
Container Type Canister
Density 1.4 g/cm3 (20°C)
Description 310 x 230 x 180 mm
For Use With (Application) Removes Stubborn Oily Residues and Other Organic Residues
Volume (Metric) 10 L
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • neodisher LaboClean FLA
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: potassium hydroxide