Learn More
Dr.Weigert™ Neodisher LaboClean F
Product Code. 11733733
Change view
Click to view available options
Volume (Metric):
12 kg
Unit Size:
12kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11733733 12 kg 12kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11733733 Supplier Dr.Weigert™ Supplier No. 410581
  • ONLINE EXCLUSIVE PRICE
    1805.00 NOK
    1782.24 NOK / 12kg
    Save 22.76 NOK
    1% Off
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
Neodisher LaboClean F - Alkaline detergent powder for the automated cleaning of commonly used laboratory utensils in food, cosmetic and in industrial laboratories. Removes grease, oil, paraffin, resin, organic dyes and felt pen residues
  • Alkaline detergent, liquid concentrate
  • neodisher LaboClean F is used for cleaning in washer disinfection
  • Free of phosphates and surfactants
  • Removes oil, paraffin, resin and organic dyes residules
  • Dosage of between 2-5 mL/L
Specifications
Product Type Neodisher F
Form Powder
pH 12.6 to 13
Sterility Non-sterile
Composition Disodium metasilicate pentahydrate, sodium carbonate, sodium hydroxide, polycarboxylates
Concentration 2 to 5 g/L
Description 280 x 270 mm
Filtered Filtered
Volume (Metric) 12 kg
compliance-icons compliance-icons
Product Identifier
  • neodisher LaboClean F
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: disodium metasilicate pentahydrate, sodium carbonate, sodiumhydroxide