Learn More
NELFB Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16418-1-AP-20UL
 View more versions of this product
Product Code. 16846034
  • 1710.33 NOK / 20µL
Description
The NELFB antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NELFB. This antibody recognizes human, mouse, rat antigen. The NELFB antibody has been validated for the following applications: IF, IP, WB, ELISA analysis.
Specifications
Specifications
NELFB
Polyclonal
Unconjugated
20 μL
25920
IgG
Western Blot, Immunoprecipitation, Immunofluorescence, ELISA
200 μg/mL
Rabbit
Primary
Human, Rat, Mouse