missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Negative Control for Mouse Monoclonal Antibody (IGG1/453) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14085JF525
This item is not returnable.
View return policy
Description
Negative Control for Mouse Monoclonal Monoclonal antibody specifically detects Negative Control for Mouse Monoclonal in Nonspecies specific samples. It is validated for Flow Cytometry,ELISA,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),Immunocytochemistry/ Immunofluorescence
Specifications
| Negative Control for Mouse Monoclonal | |
| Monoclonal | |
| Janelia Fluor 525 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Non-species specific | |
| Purified |
| Flow Cytometry, ELISA, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunocytochemistry/Immunofluorescence | |
| IGG1/453 | |
| 50mM Sodium Borate | |
| Recombinant full-length human Negative Control for Mouse Monoclonal protein | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction