Learn More
Nefopam hydrochloride, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Nefopam hydrochloride (Fenazoxine hydrochloride) is a centrally-acting but non-opioid analgesic drug, for the relief of moderate to severe pain. Nefopam hydrochloride targets β-catenin protein level in mesenchymal cells in-vitro and in-vivo.
Specifications
Specifications
| Chemical Name or Material | Nefopam hydrochloride |
| CAS | 23327-57-3 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Quantity | 10 mg |
| Synonym | Fenazoxine hydrochloride |
| Solubility Information | H2O : 12.5 mg/mL (43.13 mM; Need ultrasonic) ∣DMSO : 12.5 mg/mL (43.13 mM; Need ultrasonic) |
| SMILES | CN1CCOC(C2=CC=CC=C2)C3=CC=CC=C3C1.[H]Cl |
| Molecular Weight (g/mol) | 289.8 |
| Percent Purity | 99.78% |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.