Learn More
Neflamapimod, MedChemExpress
Product Code. 16463818
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16463818 10 mg 10mg
16440197 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16463818 Supplier Medchem Express Supplier No. HY10328
MedChemExpress Neflamapimod (VX-745) is a potent, blood-brain barrier penetrant, highly selective inhibitor of p38α inhibitor with an IC50 for p38α of 10 nM and for p38β of 220 nM. Neflamapimod (VX-745) possesses anti-inflammatory activity.
Neflamapimod (VX-745) is a potent, blood-brain barrier penetrant, highly selective inhibitor of p38α inhibitor with an IC50 for p38α of 10 nM and for p38β of 220 nM. Neflamapimod (VX-745) possesses anti-inflammatory activity.
Specifications
Chemical Name or Material Neflamapimod
CAS 209410-46-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H9Cl2F2N3OS
Quantity 10 mg
Synonym VX-745
Solubility Information DMSO : 13.08 mg/mL (29.98 mM; Need ultrasonic)
SMILES O=C1N=CN(C(C=C2)=C1C3=C(Cl)C=CC=C3Cl)N=C2SC4=CC=C(F)C=C4F
Molecular Weight (g/mol) 436.26
Formula Weight 436.26
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation