Learn More
Nefiracetam, TRC
Specifications
Specifications
| CAS | 77191-36-7 |
| Molecular Formula | C14 H18 N2 O2 |
| Quantity | 10 mg |
| Synonym | DM-9384, DZL-221, N-(2,6-dimethylphenyl)-2-oxo-1-pyrrolidineacetamide |
| InChI Formula | InChI=1S/C14H18N2O2/c1-10-5-3-6-11(2)14(10)15-12(17)9-16-8-4-7-13(16)18/h3,5-6H,4,7-9H2,1-2H3,(H,15,17) |
| SMILES | Cc1cccc(C)c1NC(=O)CN2CCCC2=O |
| IUPAC Name | N-(2,6-dimethylphenyl)-2-(2-oxopyrrolidin-1-yl)acetamide |
| Molecular Weight (g/mol) | 246.3 |
| Formula Weight | 246.1368 |
| Chemical Name or Material | Nefiracetam |